missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Veolia Water Systems™ Purification Cartridge
Reliable, robust, easy-to-use general water purification cartridge and filter kits ideal for removing ionic contaminants to achieved specific water quality.
POA
Specifications
For Use With (Application) | Removes ionic contaminants to achieve specified water quality |
---|---|
Quantity | 1 Each |
Type | Cartridge |
Description
- General water purification cartridge and filter kits
- Deionization cartridge option Q
- Purification cartridge
- Removes ionic contaminants to achieve specified water quality
Specifications
Removes ionic contaminants to achieve specified water quality | |
1 Each | |
Cartridge |