missing translation for 'onlineSavingsMsg'
Learn More

Progesterone Receptor Ab-2, Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-2, Mouse Monoclonal Antibody.

£151.00 - £689.00

Specifications

Antigen Progesterone Receptor Ab-2
Clone hPRa 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Form Concentration Quantity Price Quantity & Availability  
Product Code Brand Form Concentration Quantity Price Quantity & Availability  
12640477
View Documents Promotion Details
Epredia
MS-192-P0
Concentrated 200μg/mL 100 μL
£151.00
100µL
Please to purchase this item. Need a web account? Register with us today!
12660477
View Documents Promotion Details
Epredia
MS-192-P1
Concentrated 200μg/mL 500 μL
£483.00
500µL
Please to purchase this item. Need a web account? Register with us today!
12630477
View Documents Promotion Details
Epredia
MS-192-P
Concentrated 200μg/mL 1 mL
£689.00
1mL
Please to purchase this item. Need a web account? Register with us today!
12650477
View Documents Promotion Details
Epredia
MS-192-P1ABX
Concentrated 1mg/mL 100 μL
£474.00
100µL
Please to purchase this item. Need a web account? Register with us today!
12670477
View Documents Promotion Details
Epredia
MS-192-PABX
Concentrated 1mg/mL 200 μL
£668.00
200µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 2

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Horse, Rabbit, and Chicken. Does not react with monkey. Others not known.

Epitope: N-terminal half of human progesterone

Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form)

Positive Control: T47D cells or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Progesterone Receptor Ab-2
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Mouse
Cancer and Tumor Biology
PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Primary
hPRa 2
Monoclonal
Concentrated
RUO
Bovine, Chicken, Equine, Human, Rabbit
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications