missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Personal White Seat Covers

Individually dispensed seat covers for washrooms where hygiene is of maximum value. Kimberly-Clark™ Professional Personal White Seat Covers are flushable covers that add protection and help stop the spread of germs.

Brand:  Kimberly-Clark Professional™ 6140

Code : 02

Additional Details : Weight : 0.01000kg

Product Code. 12675636

  • £342.00 / Pack of 12
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

  • Ideal for areas where maximum hygiene levels are required
  • White
  • Individually dispensed sheets
  • Can be flushed away after use
Specifications

Specifications

Personal Seat Cover Refill
38.1 cm
White
Flat folded
1500/Pk.
45.7 cm
Virgin Fiber
45.7 x 38.1 cm
Ideal for high hygiene washrooms, designed to provide added protection whilst avoiding the spread of germs, add valued enhancement and convenience to any washroom
1-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers