Learn More
Techne™ Probe Insert Block
Click to view available options
19.5 mm
Description
- Change capacity, testing type, accuracy, or portability of Techne Calibration calibrators with these additional parts
- For use with Techne High Temperature Dri-Block Calibrator
Specifications
Specifications
Material | Aluminum Bronze |
Type | Probe Insert |
Diameter (Metric) | 19.5 mm |