missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKX Recombinant Human Protein
PRKX Recombinant Human Protein
Brand: Invitrogen PV3813
Code : 50
Additional Details : Weight : 0.05250kg
Description
PRKX is a serine⁄threonine kinase related to the catalytic subunit of the cAMP-dependent protein kinases.Specifications
NP_005035 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
68.4kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
PRKX, PKX1 | |
PRKX was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 1μM cAMP, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL kemptide peptide substrate (LRRASLG) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
PRKX Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.38mg/mL | |
3960, 5613 | |
PRKX Recombinant Human Protein | |
–80°C | |
1 Each | |
PRKX Recombinant Human Protein | |
PKA | |
440nmoles of phosphate transferred to Kemptide peptide substrate (LRRASLG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL | |
GST Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.