missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PRKCI (PKC iota), His Tag Recombinant Protein
Human PRKCI (PKC iota), His Tag Recombinant Protein
Brand: Invitrogen PV3183
Code : 50
Additional Details : Weight : 3.36000kg
Description
The PKC family of serine⁄threonine kinases, including PRKCI (PKC iota), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.Specifications
NP_002731 | |
5584 | |
5 μg | |
PKC Kinase Family | |
Recombinant | |
Storage Buffer: 20mM HEPES, pH 7.5, 150mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% NP-40, 5mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: AGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
protein kinase C, iota | |
Kinases & Inhibitors | |
Serine/Threonine Kinases |
Kinase Assay | |
71.5 kDa | |
PRKCI (PKC iota) | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
PRKCI | |
Baculovirus | |
Recombinant, Full Length | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.