missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PRKACB (PRKAC beta), GST Tag Recombinant Protein
Human PRKACB (PRKAC beta), GST Tag Recombinant Protein
£431.00 - £6530.00
Specifications
Gene Alias | PKAb, cAPKb, MGC9320, MGC41879, DKFZp781I2452 |
---|---|
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489013
|
Invitrogen™
PV6388 |
10μg |
£431.00
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15499013
|
Invitrogen™
PV6389 |
100μg |
£1460.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15409023
|
Invitrogen™
PV6390 |
1mg |
£6530.00
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The PKA catalytic subunit consists of three gene products: C-alpha, C-beta, and C-gamma. The catalytic subunit C-beta has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
PKAb, cAPKb, MGC9320, MGC41879, DKFZp781I2452 | |
Human | |
Recombinant | |
GST Tag |