Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Brand:  Thermo Scientific™ 06.5204
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12756208
  • £135.10 / Each
In Stock
to see stock.
Add to basket
Description
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
Specifications
Specifications
5µm filter with hardness stabilizer
LabTower EDI, LabTower RO
Filter with Stabilizer