Learn More
Grant Instruments™ PQ Documentation
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ PQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
Brand: Grant Instruments™ PQ R4
Code : 57
Additional Details : Weight : 0.10000kg
Description
Performance Qualification (PQ): Instrument is continuing to meet its specification- Each documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
- PQ documentation can be ordered at any time.
Specifications
Grant R4 Refrigerated Baths/Circulators | |
PQ Documentation |