missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodobismuthate EU Pharmacopoeia, Fisher Chemical™

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Brand:  Fisher Chemical RP 174 100


Product Code. 12910514

  • £87.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

100 mL
EU Ph Ref: 1070600
Eu. Ph.
Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium iodobismuthate solution. European Pharmacopoeia Ref: 1070600 (RP 174)

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.

Precautionary Statement
  • P264-Wash hands, forarms and face thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P314-Get medical advice/attention if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : acetic acid, potassium iodide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers