missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide concentrate, Honeywell™
Click to view available options
Quantity:
1 Pc.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 7681-11-0 |
Quantity | 1 Pc. |
Molecular Formula | IK |
Linear Formula | KI |
MDL Number | MFCD00011405 |
UN Number | NONH for all modes of transport |
Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
SMILES | [K+].[I-] |
IUPAC Name | potassium;iodide |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'