missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, puriss. p.a., ≥86% (T), Honeywell Fluka™

Catalog Number 15695960
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
This item is not returnable. View return policy
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
[OH-].[K+]
This item is not returnable. View return policy
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
[OH-].[K+]

Puriss. p.a., pellets,≥86% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Linear Formula KOH
UN Number UN1813
Quantity 1 kg
IUPAC Name potassium;hydroxide
Formula Weight 56.11g/mol
Percent Purity ≥86% (T)
Vapor Pressure 1 mmHg (719°C)
Chemical Name or Material Potassium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'