missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, BAKER, J.T. Baker™
Click to view available options
Quantity:
5 kg
Packaging:
HDPE
Description
A grade of chemicals for routine research usually controlled by physical criteria and often by assay (by GC).
Specifications
Color | White |
Boiling Point | 1327°C |
CAS | 1310-58-3 |
Melting Point | 360°C |
Molecular Formula | HKO |
MDL Number | MFCD00003553 |
Density | 2.04g/cm3 at 20°C |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
Solubility Information | In Water: 1120g/L |
Quantity | 5 kg |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'