missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexachloroiridate(III) hydrate, Ir 32.8% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Potassium hexachloroiridate(III) hydrate is used to prepare other iridium compounds. It is used as a redox mediator to visualize human finger prints on a poly(vinylidene difluoride) membrane by scanning electrochemical microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline Powder |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 522.22 (Anhydrous) |
Concentration or Composition (by Analyte or Components) | Ir 33.4% min. |
Chemical Name or Material | Potassium hexachloroiridate(III) hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'