missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride Dihydrate, 98%, Reagent grade, Honeywell™
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Quantity | 100 g |
Linear Formula | KF · 2H2O |
UN Number | UN1812 |
IUPAC Name | potassium;fluoride;dihydrate |
Formula Weight | 94.13g/mol |
Percent Purity | 98% |
Grade | Reagent |
Packaging | Poly Bottle |
Chemical Name or Material | Potassium Fluoride Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'