missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Solution, Honeywell™ Fluka™

Product Code. 15605930
Click to view available options
Quantity:
500 mL
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

conductivity 111.8 mS/cm at 25 °C, conductance standard

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7447-40-7
Color Colorless
Physical Form Liquid
Quantity 500 mL
Molecular Formula ClK
Linear Formula KCl
MDL Number MFCD00011360
UN Number NONH for all modes of transport
Beilstein 1711999
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
Solubility Information Completely soluble in water
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium;chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.55g/mol
Packaging Plastic bottle
Density 1.04g/mL at 20°C
Chemical Name or Material Potassium chloride solution, Conductivity 111.8mS/cm at 25°C, conductance standard
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'