missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Powder, U.S.P. - F.C.C., J.T. Baker™

Product Code. 15598074
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 7447-40-7
Physical Form Powder
Quantity 2.5 kg
Molecular Formula ClK
MDL Number MFCD00011360
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium;chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.55
Grade FCC/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.99kg
Chemical Name or Material Potassium Chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'