missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, Reagent grade, Honeywell

Reagent Grade,≥98%, powder, −325 mesh

£30.72 - £898.12

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15614410
View Documents
Honeywell Research Chemicals
347825-250G
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15694400
View Documents
Honeywell Research Chemicals
347825-1KG
1 kg
£111.81
1kg
Please to purchase this item. Need a web account? Register with us today!
15604410
View Documents
Honeywell Research Chemicals
347825-2.5KG
2.5 kg
£220.47
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15684400
View Documents
Honeywell Research Chemicals
347825-12KG
12 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

584-08-7
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
[K+].[K+].[O-]C([O-])=O

Specifications

584-08-7
K2CO3
NONH for all modes of transport
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
[K+].[K+].[O-]C([O-])=O
138.21
138.21g/mol
Reagent
CK2O3
MFCD00011382
4267587
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium;carbonate
11430
≥98%
Potassium carbonate
Videos
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

missing translation for 'rtecsNumber' : TS7750000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.