missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromide, ≥99.0%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 1383°C (1013hPa) |
Melting Point | 734°C |
Color | Colorless |
pH | 5.0 to 8.0 (at 20°C) |
Physical Form | Solid |
Quantity | 500 g |
Linear Formula | KBr |
UN Number | NONH for all modes of transport |
Formula Weight | 119g/mol |
Percent Purity | ≥99.0% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'