missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Vegetable, Multi-Compendial, N.F., J.T. Baker™

£2135.00 - £12150.00

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15538304
View Documents Promotion Details
Avantor J.T.Baker
4600-04
200 mL Narrow Mouth Amber Glass Bottle
£2135.00
200mL
Please to purchase this item. Need a web account? Register with us today!
15548304
View Documents Promotion Details
Avantor J.T.Baker
4600-06
1 L Narrow Mouth Amber Glass Bottle
£12150.00
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

9005-64-5
522.68
HMFKFHLTUCJZJO-UHFFFAOYNA-N
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
MFCD00165986
443314

Specifications

9005-64-5
1L = 1.08kg
MFCD00165986
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
522.68
Multi-Compendial/NF
148.00°C
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
HMFKFHLTUCJZJO-UHFFFAOYNA-N
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
443314
Polysorbate 20, Vegetable
Videos
SDS
Documents

Documents

Product Certifications