missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multiroir™ Polypropylene Shockproof Storage Box
Guard items against damage and store them securely by using the sturdy polypropylene Multiroir™ Polypropylene Shockproof Storage Box with tight-fitting lid.
POA
Specifications
Color | Translucent |
---|---|
Material | Polypropylene |
Product Type | Storage Box |
Quantity | 1 Pack |
Description
- Sturdy and shock-resistant
- Translucent Polypropylene
Specifications
Translucent | |
Polypropylene | |
Storage Box | |
1 Pack |