Learn More
Cytiva Polypropylene On-line Filter Kit
Used for chromatography applications. Cytiva Polypropylene On-line Filter Kit are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules.
Brand: Cytiva 18-1130-23
Code : GX
Additional Details : Weight : 0.01260kg
Description
- Packsize 25 Guidance
- The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
- Polypropylene (PP)
Specifications
Polypropylene | |
Used in chromatography of biomolecules | |
1Kit | |
Online Filter Kit |