missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Nylon Filter Cloth, 300mm x 300mm
For sieving or precision filtration of biological and chemical solutions. Fisherbrand™ Nylon Filter Cloths, 300mm x 300mm are available in a comprehensive range of pore sizes.
POA
Description
- Operating temperature of -40° to +115°C
- Excellent compatibility with alcohols
- Resistant to aggressive solvents
- Applications include solvent filtration and particle analysis
Specifications
300 mm | |
Plastic |