missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Plastic divider, 41 compartment, height 75mm to fit 800mm x 750mm
Brand: Manutan A348793
Code : Z3
Dettagli aggiuntivi : Weight : 1.00000kg
Descrizione
Fully customisable drawer divider inserts.- 41 compartments
- Drawer O/A height: 75mm