missing translation for 'onlineSavingsMsg'
Learn More

Manutan Plastic divider, 41 compartment, height 100mm to fit 800mm x 750mm

Brand:  Manutan A348783

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12870819

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Fully customisable drawer divider inserts.
  • 41 compartments
  • Drawer O/A height: 100mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers