missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, 98%, Thermo Scientific Chemicals
21.38 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
PIPES is commonly used as a buffering agent in biochemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
PIPES is commonly used as a buffering agent in biochemistry.
Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
PIPES is commonly used as a buffering agent in biochemistry.
Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | >300°C |
Quantity | 25 g |
Beilstein | 817713 |
Merck Index | 147479 |
Solubility Information | Soluble in 1M sodium hydroxide solution. Slightly soluble in water. |
Formula Weight | 302.37 |
Percent Purity | 98% |
Chemical Name or Material | PIPES |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'