missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperidino(3-piperidinyl)methanone hydrochloride, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 232.75 |
Percent Purity | 90% |
Chemical Name or Material | Piperidino(3-piperidinyl)methanone hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'