missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3C2A (PI3K-C2 alpha) Recombinant Human Protein
PIK3C2A (PI3K-C2 alpha)
Brand: Invitrogen PV5586
Code : 50
Additional Details : Weight : 0.50000kg
Description
The Class II PI3 kinases, including PIK3C2A, phosphorylate the 3 position of PI or PI4P. Though not as well studied as the Class I PI3 kinases, the Class II enyzmes have been linked to diverse, recpetor-mediated cellular processes including insulin signalling, neuronal survival, and growth factor signalling.Specifications
NP_002636.1 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
PIK3C2A (PI3K-C2 alpha) Recombinant Human Protein | |
–80°C | |
1 Each | |
PIK3C2A (PI3K-C2 alpha) Recombinant Human Protein | |
PI3 | |
PIK3C2A (PI3K-C2 alpha) Recombinant Human Protein | |
Human | |
Baculovirus | |
Lipid kinase |
0.16mg/mL | |
171kDa | |
20μg | |
For research use only. Not for use in diagnostic procedures | |
PIK3C2A (PI3K-C2 alpha) | |
PIK3C2A (PI3K-C2 alpha) was diluted in assay buffer and the activity determined in a 10μL Adapta™ assay (PV5099) with 10μM ATP and 100μM PI lipid kinase substrate (PV5371) at room temperature for 1 hour. The reaction was stopped with a 5μL addition of EDTA and detection reagents. The final concentrations are 10mM EDTA, 2nM Adapta™ Eu-anti-ADP antibody and 4nM Alexa Fluor™ 647 ADP Tracer. The plate was read after a 45 min. room temperature incubation | |
Amino acids 142-1412 | |
3.74nmoles of ADP formed per minute per mg of total protein using the Adapta™ assay (PV5099). An ADP/ATP standard curve is used to calculate percent conversion of ATP to ADP. PIK3C2A (PI3K-C2 alpha) activity determined at a final protein concentration of 1250ng/mL | |
GST Tag | |
Kinase and inhibitors | |
60% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.