Learn More
Phosphoric acid, 85% aq. soln., Thermo Scientific Chemicals
£29.65 - £285.42
Chemical Identifiers
CAS | 7664-38-2 |
---|---|
Molecular Formula | H3O4P |
Molecular Weight (g/mol) | 97.994 |
MDL Number | MFCD00011340 |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico |
PubChem CID | 1004 |
ChEBI | CHEBI:26078 |
IUPAC Name | phosphoric acid |
SMILES | OP(=O)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11459216
|
Thermo Scientific Alfa Aesar
A18067.AP |
500 mL |
£29.65
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469216
|
Thermo Scientific Alfa Aesar
A18067.0F |
2500 mL |
£89.34
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479216
|
Thermo Scientific Alfa Aesar
A18067.0D |
10,000 mL |
£285.42
10000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.
Notes
Incompatible with strong bases and powdered metals.
Chemical Identifiers
7664-38-2 | |
97.994 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
1004 | |
phosphoric acid |
H3O4P | |
MFCD00011340 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
CHEBI:26078 | |
OP(=O)(O)O |
Specifications
7664-38-2 | |
85% aq. soln. | |
H3O4P | |
MFCD00011340 | |
14,7344 | |
Miscible with water,ethanol and mixture of ether: alcohol (3:1). | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
Phosphoric acid |
19°C to 21°C | |
1.7 | |
H3PO4 | |
UN1805 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
98 |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P264b-P280-P301+P330+P331-P303+P361+P353-P305+P351+P338-P310-P342+P311-P363-P390-P501c
H290-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PHOSPHORIC ACID SOLUTION
missing translation for 'einecsNumber' : 231-633-2
missing translation for 'rtecsNumber' : TB6300000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Phosphoric acid, 85% aqueous solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Orthophosphoric acid
RUO – Research Use Only