missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MCL-1 (Ser159) Monoclonal Antibody (RBCERNR), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9038-42

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15578226

  • £279.00 / 100 tests
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mcl-1 (Myeloid cell leukemia-1) is a member of the bcl-2 family. The carboxy terminal of Mcl-1 and bcl-2 share significant sequence homology. Expression of Mcl-1 is increased upon exposure of ML-1 cells to various types of DNA damaging agents (e.g. ionizing radiation, ultraviolet radiation, and alkylating drugs) along with increases in GADD45 and Bax and a decrease in bcl-2. Enhanced expression of Mcl-1, prominently associated with mitochondria, complements the continued expression of bcl-2 in ML-1 cells undergoing differentiation. Like bcl-2, Mcl-1 has the capacity to promote cell viability under conditions that otherwise cause apoptosis.

Phospho-MCL-1 (Ser159) Monoclonal antibody specifically detects Phospho-MCL-1 (Ser159) in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Phospho-MCL-1 (Ser159)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P97287, Q07820
MCL1
Affinity chromatography
RUO
17210, 4170
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
RBCERNR
APC
MCL1
AW556805; BCL2 family apoptosis regulator; BCL2L3; bcl2-L-3; Bcl-2-like protein 3; bcl-2-related protein EAT/mcl1; EAT; Induced myeloid leukemia cell differentiation protein Mcl-1; induced myeloid leukemia cell differentiation protein Mcl-1 homolog; Mcl1; Mcl-1; mcl1/EAT; MCL1-ES; MCL1L; MCL1S; MCL-1S; MGC104264; MGC1839; myeloid cell leukemia 1; myeloid cell leukemia ES; myeloid cell leukemia sequence 1; myeloid cell leukemia sequence 1 (BCL2-related); TM
Mouse
100 Tests
Primary
Human, Mouse
Antibody
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.