missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylacetyl chloride, 98%, Thermo Scientific Chemicals
£15.33 - £94.80
Chemical Identifiers
CAS | 103-80-0 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.59 |
MDL Number | MFCD00000729 |
InChI Key | VMZCDNSFRSVYKQ-UHFFFAOYSA-N |
Synonym | phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl |
PubChem CID | 7679 |
IUPAC Name | 2-phenylacetyl chloride |
SMILES | ClC(=O)CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10021470
|
Thermo Scientific Acros
130300050 |
5 g | |||||||
10143660
|
Thermo Scientific Acros
130301000 |
100 g |
£32.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10479630
|
Thermo Scientific Acros
130305000 |
500 g |
£94.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103-80-0 | |
154.59 | |
VMZCDNSFRSVYKQ-UHFFFAOYSA-N | |
7679 | |
ClC(=O)CC1=CC=CC=C1 |
C8H7ClO | |
MFCD00000729 | |
phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl | |
2-phenylacetyl chloride |
Specifications
103-80-0 | |
94°C to 95°C (12 mmHg) | |
C8H7ClO | |
MFCD00000729 | |
Solubility in water: Reacts | |
ClC(=O)CC1=CC=CC=C1 | |
154.59 | |
154.6 |
1.16 | |
102°C | |
1.532 to 1.533 | |
phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl | |
VMZCDNSFRSVYKQ-UHFFFAOYSA-N | |
2-phenylacetyl chloride | |
7679 | |
Phenylacetyl chloride 98%, 98% |
RUO – Research Use Only