missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH Combination Electrodes

£1015.00

Specifications

Type pH Combination Electrode
Temperature (Metric) Range 0°C to 100°C
View More Specs

Products 1
Product Code Brand Length (Metric) pH Range Price Quantity & Availability  
Product Code Brand Length (Metric) pH Range Price Quantity & Availability  
10702064
View Documents
Mettler Toledo™
59903232
225 mm 0 to 12
£1015.00
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

pH Combination Electrode
0°C to 100°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ pH Combination Electrodes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.