missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 4.00 Color-Coded Buffer Solution at 20°C in Bag-in-Box, Reagecon
Easy-to-use, tested, certified, NIST traceable, and shelf-stable, color-coded buffer solution.
Brand: Reagecon BPH02
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- NIST traceable
- Bag-in-box cubitainer with tap
Specifications
Red | |
4.00 ±0.01 | |
10 L |
Accuracy ± 0.01 pH units | |
Liquid | |
Bag-in-Box Container |