Learn More
Cole-Parmer™ PFA Threaded Plug
Threaded fittings offer more permanent installation
Brand: Cole-Parmer™ 06546-39
This item is not returnable.
View return policy
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Plug | |
3/8 in. | |
Ethylene Oxide (ETO) Only | |
PFA |
Clear | |
Male NPT Fitting | |
27/32 in. | |
13/16 in. |