missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, ACS reagent, Honeywell Riedel-de Haën™

Product Code. 15651660
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

ACS reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8030-30-6
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

CAS Min % ≥25.0000%
CAS Max % <50.0000%
Density 0.64 g/mL at 25 °C(lit.)
Boiling Point 40°C to 60°C
Flash Point −30°C
Packaging Stainless Steel Can
Quantity 20 L
UN Number UN3295
Grade ACS Reagent
Chemical Name or Material Petroleum ether, ACS reagent
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'