missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Pepstatin A
Click to view available options
Quantity:
5 mg
25 mg
Description
Applications: Pepstatin A, a protease inhibitor, inhibits Pepsin, Renin and Cathepsin D. Effective concentrations are 10-6 to 10-9M. It does not inhibit Trypsin, Chymotrypsin, Papain or Plasmin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
- Effective concentrations are 10-6 to 10-9M
- Does not inhibit Trypsin, Chymotrypsin, Papain or Plasmin
- Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
- MDL: MFCD00060740
Specifications
Specifications
Concentration | 10-6 to 10-9M |
Synonym | Isovaleryl-val-val-4-amino-3-hydroxy-6-methylheptanoyl-ala-4-amino-3-hydroxy-6-methylheptanoic acid SDS Certificate of Analysis Product Specification Technical Inquiry |
Solubility | Dissolves in methanol |
Beilstein | 2202362 |
Form | Powder |
InChI Key | FAXGPCHRFPCXOO-JHTFMCFMSA-N |
SMILES | CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O |
IUPAC Name | 3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid |
Molecular Weight (g/mol) | 685.90 |
Quantity | 5 mg |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'