missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDGFRA [D842V] Recombinant Human Protein
PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.
£1602.00 - £6900.00
Specifications
Accession Number | NP_006197, D842V |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 89.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15418623
|
Invitrogen™
PR7741A |
100μg |
£1602.00
100µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10671705
|
Invitrogen™
PV4292 |
1mg |
£6900.00
1mg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PDGFRA D842V is expressed in a high percentage of epithelial ovarian cancers and is implicated in the pathogenesis of acute myelogenous leukemia.
Specifications
NP_006197, D842V | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
89.7kDa | |
RUO - research use only | |
PDGFRA D842V, PDGFR alpha | |
Human | |
GST Tag | |
Frozen |