Learn More
Agilent BioTek Parallel to USB Adapter
For use with Microplate Washers and Readers
Brand:  Agilent BioTek 75135
Additional Details : Weight : 0.80000kg
Product Code. 12532747
  • £335.00 / Each
Estimated Shipment: 24-07-2024
to see stock.
Description
Description
  • Parallel to USB Adapter
  • Use to connect an ELx800, ELx808, FLx800 or uQuant to a USB-only printer
  • For use with Agilent BioTek Microplate Washers and Readers
Specifications
Specifications
Parallel to USB Adapter
ELx800, ELx808, FLx800 Fluorescence Reader