Learn More
Epredia™ Paraffin Type 1, 3, 6, 9
Find the perfect fit with four different blends of highly refined paraffin and polymers, offering a choice of infiltration times and cutting characteristics.
Brand: Epredia™ 8337
Code : A0
Additional Details : Weight : 18.14000kg
Description
- All paraffins melt between 55° and 57°C and are triple filtered to 0.5μm
- Flake form has a greater surface-to-weight ratio than pellets, resulting in a faster melting time
- Type 1 contains no polymer and offers rapid infiltration
- Type 3 contains minimal polymer and offers fast infiltration
- Type 6 is harder than Type 3 and allows for thinner sections
- Type 9 has the highest polymer content available and yields extremely thin sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable bags
Specifications
54° to 57°C | |
Paraffin | |
2 lb. | |
Infiltration and Embedding |
Safety and Handling
- Warning
Hazard Category
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H335-May cause respiratory irritation.
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P314-Get medical advice/attention if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : parafin waxes and hydrocarbon waxes, 2,6-di-tert-butyl-p-cresol, benzene,ethenylmethyl-,polymer with