Learn More
Epredia™ Paraffin Type 1, 3, 6, 9
Find the perfect fit with four different blends of highly refined paraffin and polymers, offering a choice of infiltration times and cutting characteristics.
Brand: Epredia™ 8335
Code : A0
Additional Details : Weight : 0.40000kg
Description
- All paraffins melt between 55° and 57°C and are triple filtered to 0.5μm
- Flake form has a greater surface-to-weight ratio than pellets, resulting in a faster melting time
- Type 1 contains no polymer and offers rapid infiltration
- Type 3 contains minimal polymer and offers fast infiltration
- Type 6 is harder than Type 3 and allows for thinner sections
- Type 9 has the highest polymer content available and yields extremely thin sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable bags
Specifications
54° to 57°C | |
Paraffin | |
2 lb. | |
Infiltration and Embedding |