missing translation for 'onlineSavingsMsg'
Learn More

p57 Kip2 Ab-3, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Deliver accurate, reproducible results with the Epredia™ p57Kip2 Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia MS-897-P1ABX

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12692557

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

p57Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms.

Host Species: Mouse

Clone: KP39

Isotype: IgG2b/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: Not determined

Immunogen: Recombinant human p57Kip2 protein

Molecular Weight: 57kDa

Positive Control: LS174T cells, Colon CA or placenta

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunoprecipitation (Native verified)
Specifications

Specifications

p57superscriptKip2superscript
Monoclonal
1mg/mL
Mouse
100 μL
Cell Cycle
Human, Mouse
IgG2b κ
Immunohistochemistry (Paraffin), Immunoprecipitation
KP39
Unconjugated
Recombinant human p57superscriptKip2superscript protein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers