Learn More
IKA™ RCT Digital Magnetic Stirring Hotplate
RCT digital stirrer has a round top made of aluminum alloy, stirs volumes of up to 20 L (H2O), and runs at speeds ranging from 50 to1500 rpm.
POA
Description
Description
  • Strong motor for a high range of speeds
  • Additional temperature control mode for faster heating
  • Exact temperature and speed setting via digital display
  • Set safety temperature limit displayed digitally
  • Hot top indicator
  • Digital error code display
  • Enclosed assembly guarantees long service life