missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Tolylacetylene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.
4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼21°C |
Density | 0.916 |
Boiling Point | 168°C to 170°C |
Flash Point | 49°C (120°F) |
Odor | Pungent |
Refractive Index | 1.547 |
Quantity | 5 g |
UN Number | UN3295 |
Beilstein | 969653 |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'