Learn More
Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts
Choose these replacement parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
POA
Specifications
For Use With (Equipment) | Owl™ A3-1 and A6 Wide Gel System |
---|---|
Product Line | Owl |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11952519
|
Thermo Scientific™
A3-1-EG |
Replacement end gates (1 pair) |
POA
Pack of 2 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
12444537
|
Thermo Scientific™
A3-GK-1 |
Replacement Gaskets (1 pair) |
POA
2 sets |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
12454537
|
Thermo Scientific™
A3-UVT-1 |
UVT gel tray with gasketed end gates |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
Choose Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
The gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run utilizing a wall comb. Built in buffer exchange ports allows buffer recirculation on extended runs, if needed. The large number of comb slots gives you sample and resolving distance flexibility.Specifications
Owl™ A3-1 and A6 Wide Gel System | |
Owl |