missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Run multiple gels simultaneously with Thermo Scientific™ Owl™ A2-OK Multiple Gel Electrophoresis Systems, which are also ideal for teaching labs.
Key Features:- Up to six 7 x 8cm minigels can be run on one system
- Screen up to 120 samples at once with supplied combs
- Thermo Scientific™ EasyCast™ casting design permits casting without tape or plugs
- UV-transmissible (UVT) gel trays may be placed directly on the transilluminator for easy photodocumentation
- Teacher can instruct up to 12 students using one system and one power supply
Includes:
Buffer chamber with three-point leveling base;Thermo Scientific™ SuperSafe™ lid with attached power supply leads; Six EasyCast UVT gasketed gel trays; Twelve 10-well, double-sided combs, 1.0/1.5mm thick; Two external multiple gel casters; Gel tray holder
Specifications
CE marked | |
7 x 8 cm | |
Multiple Gel Electrophoresis System | |
2.3 L | |
14.8 in. | |
8 cm | |
32.5 cm | |
4.1 in. |
Gel Size: Up to six 7 x 8cm; Buffer volume: 2.3L; L x W x H: 37.5 × 32.5 × 10.5cm | |
Owl | |
5 to 144 | |
14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm) | |
37.5 cm | |
12.8 in. | |
7 cm | |
10.5 cm |