missing translation for 'onlineSavingsMsg'
Learn More

OSGIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15248-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824344

  • £339.00 / 150µL
Estimated Shipment: 05-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

OSGIN1 regulates the differentiation and proliferation of normal cells through the regulation of cell death.
Specifications

Specifications

OSGIN1
Polyclonal
Unconjugated
OSGIN1
BDGI, huOKL38, OKL38, OSGIN1
Rabbit
Antigen Affinity Chromatography
RUO
171493, 29948, 71839
-20°C
Liquid
Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UJX0
Osgin1
OSGIN1 Fusion Protein Ag4295
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers