Learn More
Thermo Scientific™ Orion™ 9156BNWP Combination pH Electrode
Perform routine pH measurements under rugged conditions using this combination pH electrode.
Brand:  Thermo Scientific™ 9156SC
 View more versions of this product
Product Code. 10577591
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Perform routine pH measurements under rugged conditions using the Thermo Scientific™ Orion™ 9156BNWP Combination pH Electrode. It has an unbreakable body that extends beyond the pH sensing glass bulb for protection.
Includes:
  • 60ml bottle of Orion electrode fill solution, 900011
TRUSTED_SUSTAINABILITY
Specifications
Specifications
With screw cap connector
Glass Fiber
0 to 14
Combination pH Electrode
Ag/AgCl
32°F to 194°F
120 mm
12 mm
Screw Cap
Epoxy Body
0°C to 90°C
4M KCl with Ag/AgCl
0.02
4.7 in.
0.47 in.
16 mm