missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ FFAPplus GC Capillary Column

Brand:  Macherey-Nagel™ 726242.60

Additional Details : Weight : 0.35000kg

Product Code. 15220838

  • £880.00 / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.25 mm
GC Capillary Column
Optima™ FFAPplus
250°C to 260°C
1 Each
G1, G2, G38
0.4 mm
Free Carboxylic Acids Analysis, Free Carboxylic Acids
60 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers