Learn More
Thermo Scientific™ Open-Top Screw Caps
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
Brand:  Thermo Scientific™ TS-13217
Code : 16
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11577350
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
  • Fit into Reacti-Vial™ small reaction vials
  • Used with PTFE/Rubber laminated discs
Specifications
Specifications
TS-13217
Open Top Screw Cap
14 mL Screw Cap Septum Vials
14 mL
18 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.