missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Online Filter Holder

Used with classic ÄKTA systems.  Cytiva Online Filter Holder is fitted between the mixer and the injection valve.

Brand:  Cytiva 18-1112-44

Code : GX

Additional Details : Weight : 0.01700kg

Product Code. 10650839

  • £508.00 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

  • The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran.
  • Includes 1 x filter supplied with the filter holder.
  • Filterholder: Polyetheretherketone (PEEK)
  • Filter: Polypropylene (PP) 
Specifications

Specifications

Classic ÄKTA
Polypropylene
20, 50 and 100 mL
1 Pack
Classic ÄKTA
Online Filter Holder
1 x filter supplied with the filter holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers